3-Bromo-6-fluoro-2-methylpyridine ≥98%

Supplier: Matrix Scientific

024607-1G 024607-5G
101818-420EA 29.23 USD
101818-420 101818-422
3-Bromo-6-fluoro-2-methylpyridine ≥98%
3-Bromo-6-fluoro-2-methylpyridine

Hazard code(s): Irritant

Formula: C₆H₅BrFN
MDL Number: MFCD03095092
CAS Number: 375368-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR