3-Bromo-2,5-dichloropyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Bromo-2,5-dichloro-pyridine

024606-1G 024606-5G
101818-416EA 16.92 USD
101818-416 101818-418
3-Bromo-2,5-dichloropyridine ≥98%
3-Bromo-2,5-dichloropyridine

Hazard code(s): Irritant

Formula: C₅H₂BrCl₂N
MW: 226.89 g/mol
MDL Number: MFCD06659484
CAS Number: 138006-41-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR