3-Thiophenecarbonitrile ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Cyanothiophene, 3-Thienyl cyanide, Thiophene-3-carbonitrile

024604-5G 024604-25G 024604-100G
101818-408EA 24.62 USD
101818-408 101818-410 102832-920
3-Thiophenecarbonitrile ≥98%
3-Thiophenecarbonitrile

Hazard code(s): Irritant, Irritant-Harmful

Formula: C₅H₃NS
MW: 109.15 g/mol
Boiling Pt: 82 °C (10mm)
Density: 1.1956 g/cm³ (20 °C)
MDL Number: MFCD00151852
CAS Number: 1641-09-4
EINECS: 216-687-7
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 82°C/10mm
Density 1.2
ND20 1.56

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR