4-Bromo-2-thenoic acid ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 4-Bromo-2-thiophenecarboxylic acid

024601-1G 024601-5G 024601-25G
101818-402EA 13.85 USD
101818-402 101818-404 101818-406
4-Bromo-2-thenoic acid ≥98% for synthesis
4-Bromo-2-thenoic acid

Hazard code(s): Irritant

Formula: C₅H₃BrO₂S
MDL Number: MFCD03422294
CAS Number: 16694-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR