Methyl-3-hydroxybenzoate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 3-Hydroxybenzoate, 3-Hydroxybenzoic acid methyl ester

024173-25G 024173-100G 024173-500G
101817-558EA 129.23 USD
101817-558 101817-560 101817-562
Methyl-3-hydroxybenzoate ≥97%
Methyl-3-hydroxybenzoate

Hazard code(s): Irritant

Formula: C₈H₈O₃
MW: 152.15 g/mol
MDL Number: MFCD00002295
CAS Number: 19438-10-9
EINECS: 243-071-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR