3-(3,4-Dimethoxyphenyl)propionic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3,4-Dimethoxyhydrocinnamic acid, 3-(3,4-Dimethoxyphenyl)propanoic acid

023819-5G 023819-25G
101816-838EA 15.38 USD
101816-838 101816-840
3-(3,4-Dimethoxyphenyl)propionic acid ≥95%
3-(3,4-Dimethoxyphenyl)propionic acid

Hazard code(s): Irritant

Formula: C₁₁H₁₄O₄
MW: 210.23 g/mol
Boiling Pt: 344 °C (1013 hPa)
Melting Pt: 97…98 °C
MDL Number: MFCD00002774
CAS Number: 2107-70-2
EINECS: 218-288-3

Order Now

Specification Test Results

Melting point 97-98°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR