2-Amino-5-chloro-3-nitropyridine ≥98% for synthesis

Supplier: Matrix Scientific
Warning

Synonyms: 5-Chloro-3-nitro-2-pyridylamine, 5-Chloro-3-nitropyridin-2-amine

023812-25G 023812-100G
101816-816EA 44.62 USD
101816-816 101816-818
2-Amino-5-chloro-3-nitropyridine ≥98% for synthesis
2-Amino-5-chloro-3-nitropyridine

Hazard code(s): Irritant

Formula: C₅H₄ClN₃O₂
MW: 173.56
Melting Pt: 190…195 °C
MDL Number: MFCD00092011
CAS Number: 5409-39-2

Order Now

Specification Test Results

Melting point 190-195°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR