2,3-Diaminobenzoic acid ≥95%

Supplier: Matrix Scientific

023801-5G 023801-25G
101816-782EA 47.69 USD
101816-782 101816-784
2,3-Diaminobenzoic acid ≥95%
2,3-Diaminobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₈N₂O₂
Melting Pt: 198…204 °C
MDL Number: MFCD00137818
CAS Number: 603-81-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR