4-Bromo-3-(trifluoromethyl)benzenesulfonyl chloride ≥97%
Supplier: Matrix Scientific
023775-5G
023775-25G
101816-712EA
32.31
USD
101816-712
101816-714
4-Bromo-3-(trifluoromethyl)benzenesulfonyl chloride ≥97%
4-Bromo-3-(trifluoromethyl)benzenesulphonyl chloride
Hazard code(s): Corrosive, Moisture Sensitive
Formula:
C₇H₃BrClF₃O₂S Boiling Pt: 92…94 °C (0.35mm) Density: 1.833 g/cm³ (20 °C) |
MDL Number:
MFCD03094391 CAS Number: 351003-47-9 UN: 3265 ADR: 8,II |
Specification Test Results
Boiling point | 92-94°C/0.3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...