2,4-Dimethoxybenzenesulfonyl chloride ≥99%
Supplier: Matrix Scientific
023774-5G
023774-25G
023774-100G
101816-706EA
70.77
USD
101816-706
101816-708
101816-710
2,4-Dimethoxybenzenesulfonyl chloride ≥99%
2,4-Dimethoxybenzenesulphonyl chloride
Hazard code(s): Irritant
Formula:
C₈H₉ClO₄S Melting Pt: 70…73 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00060683 CAS Number: 63624-28-2 UN: 3261 ADR: 8,II |
Specification Test Results
Melting point | 70-73°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...