2,4,6-Trichlorobenzenesulfonamide ≥97%

Supplier: Matrix Scientific

023769-1G 023769-5G
101816-686EA 58.46 USD
101816-686 101816-688
2,4,6-Trichlorobenzenesulfonamide ≥97%
2,4,6-Trichlorobenzenesulphonamide

Hazard code(s): Irritant

Formula: C₆H₄Cl₃NO₂S
Melting Pt: 179…181 °C
MDL Number: MFCD00955746
CAS Number: 28460-30-2

Order Now

Specification Test Results

Melting point 179-181°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR