2-Chloro-6-methylbenzamide ≥98%

Supplier: Matrix Scientific

023755-250MG 023755-1G
101816-646EA 35.38 USD
101816-646 101816-648
2-Chloro-6-methylbenzamide ≥98%
2-Chloro-6-methylbenzamide

Hazard code(s): Irritant

Formula: C₈H₈ClNO
Melting Pt: 164…166 °C
MDL Number: MFCD00052820
CAS Number: 101080-58-4

Order Now

Specification Test Results

Melting point 164-166°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR