3-Bromo-2-thenoic acid ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 3-Bromo-2-thiophenecarboxylic acid, 3-Bromothiophene-2-carboxylic acid

023753-5G 023753-25G
101816-640EA 21.54 USD
101816-640 101816-642
3-Bromo-2-thenoic acid ≥98% for synthesis
3-Bromo-2-thenoic acid

Hazard code(s): Irritant

Formula: C₅H₃BrO₂S
Melting Pt: 195…196 °C
MDL Number: MFCD00052291
CAS Number: 7311-64-0

Order Now

Specification Test Results

Melting point 195-196°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR