2-Bromo-3-fluoropyridine ≥98%

Supplier: Matrix Scientific

023741-5G 023741-25G
101816-612EA 44.62 USD
101816-612 101816-614
2-Bromo-3-fluoropyridine ≥98%
2-Bromo-3-fluoropyridine

Hazard code(s): Irritant

Formula: C₅H₃BrFN
MDL Number: MFCD04114128
CAS Number: 40273-45-8
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR