3-Formyl-6-methoxypyridine ≥97%

Supplier: Matrix Scientific

Synonyms: 6-Methoxypyridine-3-carbaldehyde, 6-Methoxynicotinaldehyde, 6-Methoxypyridine-3-carboxaldehyde, 2-Methoxypyridine-5-carboxaldehyde, 6-Methoxy-3-pyridinecarboxaldehyde

023736-1G 023736-5G
101816-598EA 15.38 USD
101816-598 101816-600
3-Formyl-6-methoxypyridine ≥97%
3-Formyl-6-methoxypyridine

Hazard code(s): Irritant

Formula: C₇H₇NO₂
MW: 137.14 g/mol
MDL Number: MFCD02683446
CAS Number: 65873-72-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR