4-Methoxy-o-toluidine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Methoxy-2-methylaniline, m-Cresidine, 2-Methyl-p-anisidine, 1-Amino-4-methoxy-2-methylbenzene, 2-Amino-5-methoxytoluene, 4-Amino-3-methylanisole

022815-25G 022815-100G
101814-786EA 40 USD
101814-786 101814-788
4-Methoxy-o-toluidine ≥95%
4-Methoxy-o-toluidine

Hazard code(s): Toxic

Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 248…249 °C
Melting Pt: 13…14 °C
Density: 1.08 g/cm³
MDL Number: MFCD00007735
CAS Number: 102-50-1
EINECS: 203-036-7
UN: 2810
ADR: 6.1 III

Order Now

Specification Test Results

Boiling point 248-249°C
Density 1.08


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR