2-Phenylpiperidine hydrochloride

Supplier: Matrix Scientific

Synonyms: 2-phenyl-piperidine hydrochloride salt

021680-1G 021680-5G
101812-536EA 241.54 USD
101812-536 101812-538
2-Phenylpiperidine hydrochloride
2-Phenylpiperidine hydrochloride
Formula: C₁₁H₁₆ClN
MW: 197.7 g/mol
MDL Number: MFCD02179093
CAS Number: 3466-81-7
EINECS: 222-422-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR