5-Bromo-2-chloro-3-nitropyridine ≥98%

Supplier: Matrix Scientific

021438-5G 021438-25G 021438-100G
101812-108EA 21.54 USD
101812-108 101812-110 101812-112
5-Bromo-2-chloro-3-nitropyridine ≥98%
5-Bromo-2-chloro-3-nitropyridine

Hazard code(s): Irritant, Irritant-Harmful

Formula: C₅H₂BrClN₂O₂
MW: 237.44 g/mol
Melting Pt: 69…72 °C
MDL Number: MFCD00222270
CAS Number: 67443-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR