3,4-Bis(trifluoromethyl)pyridine ≥97% for synthesis

Supplier: Matrix Scientific

021430-250MG 021430-1G
101812-082EA 47.69 USD
101812-082 101812-084
3,4-Bis(trifluoromethyl)pyridine ≥97% for synthesis
3,4-Bis(trifluoromethyl)pyridine

Hazard code(s): Irritant, Moisture Sensitive

Formula: C₇H₃F₆N
Boiling Pt: 42…44 °C (24mm)
MDL Number: MFCD08686956
CAS Number: 20857-46-9

Order Now

Specification Test Results

Boiling point 42-44°C/24m


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR