4-Bromo-2-chlorobenzotrifluoride ≥98%

Supplier: Matrix Scientific

021423-5G 021423-25G
101812-060EA 16.92 USD
101812-060 101812-062
4-Bromo-2-chlorobenzotrifluoride ≥98%
4-Bromo-2-chlorobenzotrifluoride

Hazard code(s): Irritant, Irritant-Harmful

Formula: C₇H₃BrClF₃
MW: 259.45 g/mol
MDL Number: MFCD08459292
CAS Number: 467435-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR