4-Bromophenylhydrazinium chloride ≥98%

Supplier: Matrix Scientific

Synonyms: (4-Bromophenyl)hydrazine hydrochloride, 4-Bromophenylhydrazine hydrochloride, 1-Bromo-4-hydrazinobenzene hydrochloride, 1-(4-Bromophenyl)hydrazine hydrochloride

021420-5G 021420-25G
101812-050EA 16.92 USD
101812-050 101812-052
4-Bromophenylhydrazinium chloride ≥98%
4-Bromophenylhydrazinium chloride

Hazard code(s): Irritant

Formula: C₆H₈BrClN₂
MW: 223.50 g/mol
Melting Pt: 213…217 °C
MDL Number: MFCD00012941
CAS Number: 622-88-8

Order Now

Specification Test Results

Melting point 213-217°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR