4-Nitrophenylhydrazine hydrochloride ≥98% for synthesis

Supplier: Matrix Scientific
Warning

Synonyms: 1-(4-Nitrophenyl)hydrazine hydrochloride, (4-Nitrophenyl)hydrazine hydrochloride, 4-Nitrophenylhydrazinium chloride

021419-25G 021419-100G
101812-046EA 18.46 USD
101812-046 101812-048
4-Nitrophenylhydrazine hydrochloride ≥98% for synthesis
4-Nitrophenylhydrazine hydrochloride
Formula: C₆H₇N₃O₂·HCl
MW: 189.6 g/mol
Melting Pt: 205…207 °C
MDL Number: MFCD00040592
CAS Number: 636-99-7
EINECS: 211-273-2
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Melting point 205-207°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR