5-Bromo-2-chloro-3-methylpyridine ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 5-Bromo-2-chloro-3-picoline

021415-5G 021415-25G
101812-034EA 29.23 USD
101812-034 101812-036
5-Bromo-2-chloro-3-methylpyridine ≥98% for synthesis
5-Bromo-2-chloro-3-methylpyridine

Hazard code(s): Irritant, Refrigerate

Formula: C₆H₅BrClN
MW: 206.47
Melting Pt: 40…44 °C
MDL Number: MFCD03095093
CAS Number: 29241-60-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR