3-Bromo-5-formylpyridine ≥96%

Supplier: Matrix Scientific

021413-1G 021413-5G
101812-030EA 20 USD
101812-030 101812-032
3-Bromo-5-formylpyridine ≥96%
3-Bromo-5-formylpyridine

Hazard code(s): Irritant, Refrigerate

Formula: C₆H₄BrNO
Melting Pt: 98…102 °C
MDL Number: MFCD03265758
CAS Number: 113118-81-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR