4-(Trifluoromethyl)phenylhydrazinium chloride ≥97%

Supplier: Matrix Scientific

Synonyms: (4-(Trifluoromethyl)phenyl)hydrazine hydrochloride, 4-(Trifluoromethyl)phenylhydrazine hydrochloride, 4-Hydrazinobenzotrifluoride hydrochloride, 1-(4-(Trifluoromethyl)phenyl)hydrazine hydrochloride

021412-5G 021412-25G
101812-026EA 18.46 USD
101812-026 101812-028
4-(Trifluoromethyl)phenylhydrazinium chloride ≥97%
4-(Trifluoromethyl)phenylhydrazinium chloride

Hazard code(s): Irritant, Refrigerate

Formula: C₇H₈ClF₃N₂
MW: 212.6 g/mol
MDL Number: MFCD00204233
CAS Number: 2923-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR