2-Fluoronicotinonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Cyano-2-fluoropyridine

021411-1G 021411-5G 021411-25G
101812-020EA 15.38 USD
101812-020 101812-022 101812-024
2-Fluoronicotinonitrile ≥95%
2-Fluoronicotinonitrile

Hazard code(s): Irritant, Irritant-Harmful, Store in Freezer

Formula: C₆H₃FN₂
MW: 122.1 g/mol
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: 27…30 °C
MDL Number: MFCD03095082
CAS Number: 3939-13-7

Order Now

Specification Test Results

Melting point 27-30°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR