5'-Bromo-2'-hydroxyacetophenone ≥98%

Supplier: Matrix Scientific

Synonyms: 1-(5-Bromo-2-hydroxyphenyl)ethan-1-one, 1-(5-Bromo-2-hydroxyphenyl)ethanone

021409-25G 021409-100G
101812-012EA 46.15 USD
101812-012 101812-014
5'-Bromo-2'-hydroxyacetophenone ≥98%
5'-Bromo-2'-hydroxyacetophenone

Hazard code(s): Irritant

Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Melting Pt: 58…61 °C
MDL Number: MFCD00191850
CAS Number: 1450-75-5

Order Now

Specification Test Results

Melting point 58-61°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR