2-Amino-1,3-propanediol ≥97%

Supplier: Matrix Scientific

Synonyms: Serinol, 2-Amino-1,3-dihydroxy propane, 2-Aminopropane-1,3-diol

021408-25G 021408-100G
101812-008EA 29.23 USD
101812-008 101812-010
2-Amino-1,3-propanediol ≥97%
2-Amino-1,3-propanediol

Hazard code(s): Irritant, Moisture Sensitive

Formula: C₃H₉NO₂
MW: 91.11 g/mol
Boiling Pt: 267 °C (1013 hPa)
Melting Pt: 91 - 95 °C
MDL Number: MFCD00051487
CAS Number: 534-03-2
EINECS: 208-584-0

Order Now

Specification Test Results

Boiling point 128°C/0.8mm
Melting point 55-57°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR