4-Chloro-m-toluidine ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 4-Chloro-3-methylaniline, 5-Amino-2-chlorotoluene

021407-25G 021407-100G
101812-004EA 87.69 USD
101812-004 102551-022
4-Chloro-m-toluidine ≥98%
4-Chloro-m-toluidine

Hazard code(s): Irritant, Irritant-Harmful

Formula: C₇H₈ClN
MW: 141.6 g/mol
Boiling Pt: 241 °C (1013 hPa)
Melting Pt: 83…84 °C
MDL Number: MFCD00066332
CAS Number: 7149-75-9
UN: 2239
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 83-84°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR