7-Azaindole ≥98%

Supplier: Matrix Scientific

Synonyms: 1H-Pyrrolo[2,3-b]pyridine

021403-25G 021403-100G
101811-988EA 86.15 USD
101811-988 102551-020
7-Azaindole ≥98%
7-Azaindole

Hazard code(s): Irritant

Formula: C₇H₆N₂
MW: 118.14 g/mol
Boiling Pt: 384 °C (1013 hPa)
Melting Pt: 103 - 106 °C
Density: 1.301 g/cm³ (20 °C)
MDL Number: MFCD00005606
CAS Number: 271-63-6
EINECS: 205-981-0

Order Now

Specification Test Results

Melting point 105-107°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR