2-Bromo-3-fluoroanisole ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-1-fluoro-3-methoxybenzene

021402-5G 021402-25G
101811-982EA 40 USD
101811-982 101811-984
2-Bromo-3-fluoroanisole ≥98%
2-Bromo-3-fluoroanisole

Hazard code(s): Irritant

Formula: C₇H₆BrFO
MW: 205.03 g/mol
Boiling Pt: 220 °C (755mm Hg)
MDL Number: MFCD03788540
CAS Number: 446-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR