4,5-Difluoro-2-nitrobenzoic acid ≥95%
Supplier: Matrix Scientific
021400-5G
021400-25G
101811-978EA
26.15
USD
101811-978
101811-980
4,5-Difluoro-2-nitrobenzoic acid ≥95%
4,5-Difluoro-2-nitrobenzoic acid
Hazard code(s): Irritant
Formula:
C₇H₃F₂NO₄ |
MDL Number:
MFCD00799521 CAS Number: 20372-63-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...