3-Fluoro-4-hydroxybenzonitrile ≥98% for synthesis

Supplier: Matrix Scientific

021398-25G 021398-100G
101811-972EA 80 USD
101811-972 102663-634
3-Fluoro-4-hydroxybenzonitrile ≥98% for synthesis
3-Fluoro-4-hydroxybenzonitrile

Hazard code(s): Irritant

Formula: C₇H₄FNO
MDL Number: MFCD00215834
CAS Number: 405-04-9
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR