5-Chloro-2-fluorobenzenesulfonyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Chloro-2-fluorobenzene-1-sulphonyl chloride

021393-5G 021393-25G
101811-950EA 40 USD
101811-950 101811-952
5-Chloro-2-fluorobenzenesulfonyl chloride ≥97%
5-Chloro-2-fluorobenzenesulphonyl chloride

Hazard code(s): Irritant

Formula: C₆H₃Cl₂FO₂S
MW: 229.06 g/mol
Melting Pt: 41…44 °C
MDL Number: MFCD03094221
CAS Number: 351003-49-1
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Melting point 41-44°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR