(±)-1,2-Octanediol ≥98%
Supplier: Matrix Scientific
Synonyms:
1,2-Octanediol, (±)-1,2-Octylene glycol, (±)-1,2-Dihydroxy octane
021387-25G
021387-100G
101811-932EA
38.46
USD
101811-932
101811-934
(±)-1,2-Octanediol ≥98%
(±)-1,2-Octanediol
Hazard code(s): Irritant, Refrigerate
Formula:
C₈H₁₈O₂ MW: 146.23 g/mol Boiling Pt: 131…133 °C (10mm) Melting Pt: 36…38 °C |
MDL Number:
MFCD00010738 CAS Number: 1117-86-8 EINECS: 214-254-7 |
Specification Test Results
Boiling point | 131-133°C/1 |
Melting point | 36-38°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...