3-Bromo-5-fluoroanisole ≥98%
Supplier: Matrix Scientific
021385-25G
021385-100G
101811-924EA
73.85
USD
101811-924
101811-926
3-Bromo-5-fluoroanisole ≥98%
3-Bromo-5-fluoroanisole
Hazard code(s): Irritant
Formula:
C₇H₆BrFO Boiling Pt: 94 °C (17mm Hg) Density: 1.59 g/cm³ |
MDL Number:
MFCD01861129 CAS Number: 29578-39-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...