5-Chloronicotinic acid ≥95%
Supplier: Matrix Scientific
021377-1G
021377-5G
101811-906EA
38.46
USD
101811-906
101811-908
5-Chloronicotinic acid ≥95%
5-Chloronicotinic acid
Hazard code(s): Irritant
Formula:
C₆H₄ClNO₂ |
MDL Number:
MFCD00128901 CAS Number: 22620-27-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...