3-Amino-1-methyl-1H-pyrazole ≥98%

Supplier: Matrix Scientific

Synonyms: 1-Methyl-1H-pyrazol-3-amine, 3-Amino-1-methylpyrazole, 1-Methyl-1H-pyrazol-3-ylamine, 3-Amino-1-methyl-1H-pyrazole

021376-5G 021376-25G
101811-902EA 64.62 USD
101811-902 101811-904
3-Amino-1-methyl-1H-pyrazole ≥98%
3-Amino-1-methyl-1H-pyrazole

Hazard code(s): Corrosive, Air Sensitive

Formula: C₄H₇N₃
MW: 97.12 g/mol
Boiling Pt: 90…92 °C (1mm)
MDL Number: MFCD00466340
CAS Number: 1904-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR