5-Bromo-2-fluoro-3-methylpyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-2-fluoro-3-picoline

021373-1G 021373-5G 021373-25G
101811-886EA 15.38 USD
101811-886 101811-888 101811-890
5-Bromo-2-fluoro-3-methylpyridine ≥98%
5-Bromo-2-fluoro-3-methylpyridine

Hazard code(s): Irritant

Formula: C₆H₅BrFN
MW: 190.01 g/mol
MDL Number: MFCD03095091
CAS Number: 29312-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR