2-Bromo-6-nitrophenol ≥97%
Supplier: Matrix Scientific
021362-5G
021362-25G
101811-848EA
40
USD
101811-848
101811-850
2-Bromo-6-nitrophenol ≥97%
2-Bromo-6-nitrophenol
Hazard code(s): Irritant, Store in Freezer
Formula:
C₆H₄BrNO₃ MW: 218.01 g/mol |
MDL Number:
MFCD02683216 CAS Number: 13073-25-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...