2-Fluoro-5-iodobenzoic acid ≥97%

Supplier: Matrix Scientific
Warning

021153-5G 021153-25G
101811-460EA 67.69 USD
101811-460 101811-462
2-Fluoro-5-iodobenzoic acid ≥97%
2-Fluoro-5-iodobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₄FIO₂
MW: 266.01 g/mol
Melting Pt: 164…168 °C
MDL Number: MFCD03094517
CAS Number: 124700-41-0

Order Now

Specification Test Results

Melting point 164-168°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR