2-Chloro-4-nitropyridine ≥98%
Supplier: Matrix Scientific
021127-5G
021127-25G
101811-398EA
41.54
USD
101811-398
101811-400
2-Chloro-4-nitropyridine ≥98%
2-Chloro-4-nitropyridine
Hazard code(s): Irritant, Store in Freezer
Formula:
C₅H₃ClN₂O₂ MW: 158.54 g/mol Melting Pt: 53.5 - 54 °C |
MDL Number:
MFCD00661454 CAS Number: 23056-36-2 UN: 2811 ADR: 6.1,III |
Specification Test Results
Melting point | 50-51°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...