3-Bromopicolinic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Bromopyridine-2-carboxylic acid, 3-Bromo-2-pyridinecarboxylic acid

021126-5G 021126-25G
101811-392EA 80 USD
101811-392 101811-394
3-Bromopicolinic acid ≥97%
3-Bromopicolinic acid

Hazard code(s): Irritant

Formula: C₆H₄BrNO₂
MW: 202.01 g/mol
Melting Pt: 141…144 °C
Density: 1.813 g/cm³
MDL Number: MFCD01320380
CAS Number: 30683-23-9

Order Now

Specification Test Results

Melting point 141-144
Density 1.813

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR