2-Acetyl-3-bromothiophene ≥97%

Supplier: Matrix Scientific

021125-1G 021125-5G 021125-25G
101811-386EA 21.54 USD
101811-386 101811-388 101811-390
2-Acetyl-3-bromothiophene ≥97%
2-Acetyl-3-bromothiophene

Hazard code(s): Irritant, Light Sensitive

Formula: C₆H₅BrOS
Boiling Pt: 102…105 °C (4mm)
Melting Pt: 41…45 °C
MDL Number: MFCD00794287
CAS Number: 42877-08-7

Order Now

Specification Test Results

Boiling point 102-105°C/4
ND20 1.6108


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR