2-Amino-5-chloropyridine ≥98%
Supplier: Matrix Scientific
021118-100G
021118-500G
101811-366EA
44.62
USD
101811-366
101811-368
2-Amino-5-chloropyridine ≥98%
2-Amino-5-chloropyridine
Hazard code(s): Irritant
Formula:
C₅H₅ClN₂ Boiling Pt: 127…129 °C (21mm) Melting Pt: 135…138 °C Density: 1.514 g/cm³ (20 °C) |
MDL Number:
MFCD00006324 CAS Number: 1072-98-6 EINECS: 214-020-4 |
Specification Test Results
Boiling point | 127-129°C/2 |
Melting point | 135-138°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...