2-Bromo-5-(trifluoromethyl)benzonitrile ≥98%

Supplier: Matrix Scientific

021044-5G 021044-25G 021044-100G
101811-250EA 21.54 USD
101811-250 101811-252 101811-254
2-Bromo-5-(trifluoromethyl)benzonitrile ≥98%
2-Bromo-5-(trifluoromethyl)benzonitrile

Hazard code(s): Irritant

Formula: C₈H₃BrF₃N
MDL Number: MFCD03412187
CAS Number: 1483-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR