5-Bromo-2-fluorophenol ≥97%

Supplier: Matrix Scientific

021042-5G 021042-25G 021042-100G
101811-244EA 15.38 USD
101811-244 101811-246 101811-248
5-Bromo-2-fluorophenol ≥97%
5-Bromo-2-fluorophenol

Hazard code(s): Irritant, Store in Freezer

Formula: C₆H₄BrFO
MW: 191 g/mol
MDL Number: MFCD07782066
CAS Number: 112204-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR