(S)-(+)-2-Amino-3-methyl-1-butanol ≥97%

Supplier: Matrix Scientific

021035-25G 021035-100G
101811-222EA 149.23 USD
101811-222 101811-224
(S)-(+)-2-Amino-3-methyl-1-butanol ≥97%
(S)-(+)-2-Amino-3-methyl-1-butanol

Hazard code(s): Irritant

Formula: C₅H₁₃NO
Boiling Pt: 80…81 °C (8mm)
Melting Pt: 30…35 °C
Density: 0.92 g/cm³ (20 °C)
MDL Number: MFCD00064296
CAS Number: 2026-48-4
EINECS: 217-975-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR