Indole-5-carbaldehyd ≥98%

Supplier: Matrix Scientific

Synonyms: Indole-5-carboxaldehyde, 1H-Indole-5-carboxaldehyde

020964-5G 020964-25G
101811-112EA 49.23 USD
101811-112 101811-114
Indole-5-carbaldehyd ≥98%
Indole-5-carbaldehyd

Hazard code(s): Irritant, Store in Freezer

Formula: C₉H₇NO
MW: 145.16 g/mol
Melting Pt: 100…101 °C
MDL Number: MFCD02093664
CAS Number: 1196-69-6

Order Now

Specification Test Results

Melting point 100-101°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR