4-(Trifluoromethyl)anthranilic acid ≥96%

Supplier: Matrix Scientific

020963-5G 020963-25G 020963-100G
101811-106EA 27.69 USD
101811-106 101811-108 101811-110
4-(Trifluoromethyl)anthranilic acid ≥96%
4-(Trifluoromethyl)anthranilic acid

Hazard code(s): Irritant

Formula: C₈H₆F₃NO₂
Melting Pt: 175…176 °C
MDL Number: MFCD00519334
CAS Number: 402-13-1
EINECS: 206-937-3

Order Now

Specification Test Results

Melting point 175-176°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR